Learn More
Chemglass Life Sciences Vial, 0.3mL, 6x32mm, Clear, Round Bottom
SDP
Supplier:  Chemglass Life Sciences CV15000632
Encompass_Preferred
Vial, 0.3mL, 6x32mm, Clear, Round Bottom. These vials are used primarily with Perkin-Elmer™ ISS-100 or ISS-200 with the use of a support sleeve. Manufactured from Type 1, Class A borosilicate glass. Round bottom.
Catalog No. 50-230-859