Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 300ul, Fused Insert, Snap Top, Clear Deactivated Autosampler Vials with Write on Patch and fill lines and 100 EA matching Snap Caps with White Silicone Rubber / Tan PTFE Pre-Slit Septa fitted in the Blue Caps. 1/PK.
Supplier:  Microsolv Technology Corporation 9532C3DCP
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
Catalog No. 50-227-0627