Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 700ul, Snap Top, Clear polypropylene LCMS Compatible Autosampler Vials and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit Septa fitted in the Blue Caps. 1/PK.
Supplier:  Microsolv Technology Corporation 9572C3PC
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use. This is one of our Easy Purchase Packs.
Catalog No. 50-227-0142