Learn More
Chemglass Life Sciences Vial, Drosophila, 25 x 95mm, Narrow
SDP
Supplier:  Chemglass Life Sciences CGE4789001
Encompass
• Made from premium grade, transparent polystyrene• Narrow, wide drosophila vials are glass-like clarity for sample observation• Disposable vials are a safe and cost-effective alternative to glass vials• Narrow vials 25x95mm or wide vials 28.5x95mm available• Compatible with CGE-4790 Plugs (Sold Separately)
Catalog No. 50-189-7634