Learn More
Chemglass Life Sciences Vial, G.C. Collection, 0.1mL
SDP
Supplier:  Chemglass Life Sciences MW8101
Encompass_Preferred
Vial, G.C. Collection, 0.1mL. Vials are supplied complete with Viton o-ring (MW-100-01), PTFE faced septa (MW-104-11) and open-top screw cap (MW-102-11).
Catalog No. 50-153-2705