Learn More
Waters Corp Waters Vial Holder, 48-Well, 2 mL
Supplier:  Waters Corp 700011047
Encompass_Preferred
This 2 mL vial holder has a capacity for 48 vials. It allows samples to be prepared in advance and is compatible with ACQUITY Sample Managers.
  • Has a capacity for 48 vials.
  • Designed for 2 mL vials.
  • Compatible with ACQUITY Sample Managers, ACQUITY Arc HPLC System, and ACQUITY UPLC systems.
  • Allows samples to be prepared in advance.
  • Provided as 1 per package.
Catalog No. 50-138-5071