Learn More
Research Products International Corp Vial Rack, Polypropylene, for 20ml Vials, 50 Vial Capacity, 2 per Package
SDP
Supplier:  Research Products International Corp 224806
Encompass
Heavy duty polypropylene racks have excellent chemical resistance and can be autoclaved. Ideal for use when processing samples or transporting samples.
Catalog No. 50-550-233