Learn More
Electron Microscopy Sciences CryoELITE Benchmate Vial Rack
SDP
Supplier:  Electron Microscopy Sciences 62021
Encompass
CryoELITE Benchmate vial rack is compatible with both freestanding and round bottom vials and holds 50 cryogenic vials. Features one hand cap removal of freestanding vials. Manufactured from polypropylene. Easily cleaned in an automatic washer and autoclavable at 121 degree C for 20 minutes. Non-skid feet offer stability for bench work. Stackable. Number of wells: 5 deep x 10 wide.
Catalog No. 50-192-9352