Learn More
Cayman Chemical Vialinin A Natural Products Ex
SDP
Supplier:  Cayman Chemical 100105191 MG
Encompass_Preferred
A natural compound with strong antioxidant activity; potently inhibits the release of TNF-α (IC50 = 0.09 nM) and IL-4 (IC50 = 2.8 nM), as well as β-hexosaminidase and CCL2 (MCP-1) from IgE-stimulated RBL-2H3 mast cells
Catalog No. 50-288-9557