Learn More
Microsolv Technology Corporation Vials, EPA Style, Glass. Amber, 40ml. 95x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. 100/PK.
Supplier:  Microsolv Technology Corporation 970400AV
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Pharmaceutical grade Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Fill volume is approximately 42.9ml. Use 24mm screw caps that have 24-400 threads, sold separately.
Catalog No. 50-227-1370