Learn More
Microsolv Technology Corporation Vials, EPA Style, Glass. Clear, 40ml. 95x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. 100/PK.
Supplier:  Microsolv Technology Corporation 970410CV
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Fill volume is approximately 43ml. Use 24mm screw caps that have 24-414 threads, sold separately. Lower cost for budget minded laboratories.
Catalog No. 50-227-1074