Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear, 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Rounded Bottom with a Tapered Neck. CTC Compliant. 100/PK.
Supplier:  Microsolv Technology Corporation 95025ECT20C
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with CTC Pal, and other Headspace autosamplers.
Catalog No. 50-227-0163