Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 2ml, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Supplier:  Microsolv Technology Corporation 9502SE0DAV
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Amber vials are often used for light sensitive compounds and considered low actinic.
Catalog No. 50-227-1078