Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 1.8ml, MRQ Style. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Supplier: Microsolv Technology Corporation 9512C00DCV
First hydrolytic, Class A, Type-N51A, Deactivated / Silanized, Borosilicate Glass. The MRQ Style Vials have a fill volume of 1.2ml and a very low residual volume of 5ul due to the center-draining effects and the conical bottom with a well that is slightly larger than autosampler vial needles. Side entry port needles will recover the maximum amount during use.