Learn More
Medchemexpress LLC Viaminate | 53839-71-7 | 447.61 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112077100MG
Encompass_Preferred
Viaminate is a derivative of Retinoic acid, used for studying severe acne and other keratinization disorders. It is an amide compound formed by Tretinoin and Ethyl p-aminobenzoate, where the end group of Tretinoin is transformed into amide and ester chain.
  • Used for studying severe acne and other keratinization disorders.
  • Targets RAR/RXR.
  • Involved in metabolic enzyme/protease pathway.
  • Involved in vitamin D related/nuclear receptor pathway.
Catalog No. 50-004-38993