Learn More
Medchemexpress LLC Viaminate | 53839-71-7 | 96.74% | 447.61 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY112077A10MG
Encompass_Preferred
Viaminate is a derivative of Retinoic acid, utilized for research into severe acne and other keratinization disorders. It is an amide compound resulting from the reaction of Tretinoin and Ethyl p-aminobenzoate, where the end group of Tretinoin is converted into an amide and ester chain.
Catalog No. 50-004-39018