Learn More
Grainger Vibration Isolator Pads
Supplier:  Grainger 36VZ59.2053612776
Encompass_Preferred
The Grainger vibration isolator pads are made of neoprene rubber and provide vibration damping and isolation for industrial equipment. They have a maximum load capacity of 250 lbs and measure 3" x 3" x 7/8" thick. The neoprene pads effectively absorb vibration and impact from equipment and machinery to reduce noise and movement. Their rugged construction and durability make them well-suited for isolating vibration in industrial environments. The vibration pads can be used under heavy equipment, appliances, machinery and more to prevent noise and movement transfer. Overall, these neoprene isolator pads from Grainger offer an effective vibration damping solution for industrial applications.
Catalog No. NC2258545