Learn More
Cole-Parmer Monarch 6400-011 Vibration Meter with Sensor Pk
Supplier:  Cole-Parmer 9883403
Encompass
Use the vibration meter and electronic stethescope in combination with the accelerometer vibration sensor and headphones to quickly locate noise and chatter in your machinery. Vibration severity chart on front of meter provides instant status by relating the digital reading to machinery applications per ISO 10816-1. Software lets you store, compare, and graph your measurements. Software System Requirements: System: 486 computer or better, CD-ROM, running Windows 95/98, XP and NT 4.0; Memory: 8 MB RAM; Hard disk space: 100 MB minimum
Catalog No. 50-631-703