Learn More
Medchemexpress LLC HY-N2165 5mg Medchemexpress, Vicenin 2 CAS:23666-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21655MG
Encompass_Preferred
Medchemexpress, HY-N2165 5mg Vicenin 2 CAS:23666-13-9 Vicenin 2 is an angiotensin-converting enzyme (ACE) inhibitor (IC50=43.83 μM) from the aerial parts of Desmodium styracifolium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2871