Learn More
Medchemexpress LLC Vicenin 3 | 59914-91-9 | MFCD21333257 | 99.9% | 564.49 g·mol⁻1 | C26H28O14 | 10 MG

Supplier: Medchemexpress LLC HYN409010MG
Vicenin 3 is a high-purity flavonoid C-glycoside supplied as a small-scale research standard suitable for biochemical, pharmacological, and analytical studies. It is commonly used as an analytical reference and in assays investigating angiotensin-converting enzyme (ACE) activity.
- High purity: 99.9% suitable for analytical applications.
- Defined identity: CAS 59914-91-9 and formula C26H28O14.
- Convenient pack size: 10 MG for preliminary experiments and method development.
- Solid form: easy storage and handling under standard laboratory conditions.
- Applicable to biochemical assays and ACE inhibition studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.