Learn More
Medchemexpress LLC HY-B0277 100mg Medchemexpress, Vidarabine CAS:5536-17-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0277100MG
Encompass_Preferred
Medchemexpress, HY-B0277 100mg Vidarabine CAS:5536-17-4 Vidarabine (Ara-A) an antiviral drug which is active against herpes simplex and varicella zoster viruses[2]. Vidarabine has IC50s of 9.3 μg/ml for HSV-1 and 11.3 μg/ml for HSV-2[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1051
 Restricted Item