Learn More
Medchemexpress LLC Vidarabine monohydrate | 24356-66-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6666100MG
Encompass_Preferred
Vidarabine monohydrate is an adenine arabinoside and an antiviral agent. It is active against herpes simplex viruses (HSV) and varicella zoster viruses. This product is for research use only and not sold to patients.
  • Antiviral agent
  • Active against herpes simplex viruses (HSV)
  • Active against varicella zoster viruses
  • Soluble in DMSO (250 mg/mL)
  • For research use only
Catalog No. 50-004-20219