Learn More
Revvity Health Sciences Inc ViewPlate-96 Black, Glass Bottom, Tissue Culture Treated, Sterile, 96-Well with Lid, Case of 40
Supplier:  Revvity Health Sciences Inc 6005430
Encompass
Our ViewPlate microplates are engineered for bottom reading systems and high content imaging. Glass exhibits better optical properties and is perfectly flat, but is less-suited for cell culture. Glass therefore usually has to be coated with proteins that enhance cell adherence and growth (e.g., poly-D-lysine or collagen). This high quality microplate can be utilized in your cell based assay for superior results. Leveraging years of assay and instrument experience in plate detection, PerkinElmer designs better microplates for better performance that guarantees better results for all PerkinElmer applications. Packaging Notes: Cases of 40 glass-bottom 96-well plates are packaged as 40 individually bagged and sealed lidded plates. Well Plate Dimensions Well format: 96-well w/Glass bottom Description Specification Number of Rows 8 Number of Columns 12 Well volume 300 µL Recommended working volume 50 µL- 200 µL Height (mm) 11.6 Length (mm) 127.75 Width (mm) 85.34 Well diameter (mm) 6 Well d
Catalog No. NC0402198