Learn More
Medchemexpress LLC Vilanterol | 503068-34-6 | MFCD18782703 | 99.3% | 486.43 | C24H33Cl2NO5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY143005MG
Encompass_Preferred
Vilanterol is a long-acting β2-adrenergic receptor agonist supplied for research use. It is used in pharmacology and receptor biology studies to investigate β2-mediated signaling, potency, and selectivity in vitro.
  • Long-acting β2-adrenergic receptor agonist for pharmacology studies.
  • High purity (99.3%) suitable for analytical and biological assays.
  • Molecular formula C24H33Cl2NO5; molecular weight 486.43.
  • Available in small research quantities for assay development and pilot studies.
  • Supplied as a solid form appropriate for in vitro testing.
Catalog No. 50-002-21372