Learn More
Selleck Chemical LLC Vilanterol E4842-1G
SDP
Supplier:  Selleck Chemical LLC E48421G
Encompass_Preferred
Vilanterol (GW642444) is a potent and selective agonist of 2-adrenoceptor ( 2-AR) with pEC50 of 2-AR 1-AR and 3-AR is 10 37 6 98 and 7 36 respectively in cAMP detection assays
Catalog No. 50-000-41966