Learn More
Medchemexpress LLC Vilazodone | 163521-12-8 | MFCD09838919 | 441.5 g/mol | C26H27N5O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY142625MG
Encompass_Preferred
Vilazodone is a selective serotonin reuptake inhibitor (SSRI) and partial 5-HT1A receptor agonist supplied for research use. It is used in studies of depressive disorders, serotonergic pharmacology, and receptor binding, and is provided as a solid research compound for analytical applications.
  • Selective serotonin reuptake inhibitor and partial 5-HT1A receptor agonist.
  • Used in preclinical research of major depressive disorder and affective disorders.
  • Provided as a solid for research and analytical applications.
  • Chemical identifiers: CAS 163521-12-8; formula C26H27N5O2; molecular weight 441.5 g/mol.
  • Intended for laboratory research use only; not for human or veterinary use.
Catalog No. 50-002-21606