Learn More
Medchemexpress LLC Vilazodone (hydrochloride) | 163521-08-2 | 100.7% | C26H28ClN5O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1426110MM/1ML
Encompass_Preferred
Vilazodone hydrochloride is a serotonin transporter (SER) inhibitor and 5-HT1A receptor partial agonist. It is intended for research use only.
  • Serotonin transporter (SER) inhibitor
  • 5-HT1A receptor partial agonist
  • Attenuates stress-induced potentiated startle in rats
  • Inhibits stress-induced vocalizations in rats
  • Produces characteristic behavioral syndrome in rats
  • Represents an option for the treatment of major depressive disorder (MDD)
Catalog No. 50-003-53712