Learn More
TARGETMOL CHEMICALS INC VILAZODONE HYDROCHLORIDE 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6232200MG
Encompass_Preferred
Also available in 10 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vilazodone Hydrochloride (EMD 68843) a partial agonist of 5-HT1A receptors and specific serotonin reuptake inhibitor (SSRI) is utilized in treating of the major depressive disorder. purity: 99%
Catalog No. 50-257-6564