Learn More
Medchemexpress LLC Vildagliptin | 274901-16-5 | 99.9% | 303.40 g·mol⁻1 | C17H25N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY142915MG
Encompass_Preferred
Vildagliptin is a selective dipeptidyl peptidase-IV (DPP-IV) inhibitor supplied as a research and analytical standard. The compound (C17H25N3O2; MW 303.40 g·mol⁻1; CAS 274901-16-5) is provided in a 5 mg package with high analytical purity for use in biochemical assays, cell-based studies, and analytical method development.
  • Selective DPP-IV inhibitor for biochemical and cell-based assays.
  • High analytical purity (99.9%) suitable for quantitation and standards.
  • Well-characterized small molecule with formula C17H25N3O2 and MW 303.40 g·mol⁻1.
  • Supplied as a 5 mg research standard for method development and validation.
  • Product documentation includes SDS and COA for quality control.
Catalog No. 50-002-21573