Learn More
Apexbio Technology LLC Vildagliptin (LAF-237) 274901-16-5 50mg
SDP
Supplier:  Apexbio Technology LLC A403750
Encompass_Preferred
Vildagliptin (LAF-237 CAS 274901-16-5) is an inhibitor of dipeptidyl peptidase-4 (DPP-4) a widely expressed antigenic enzyme with roles in signal transduction apoptosis and immunoregulation Vildagliptin inhibits DPP-4 enzymatic activity at an IC50 of approximately 2 3 nM preventing the degradation of glucagon-like peptide-1 (GLP-1) Through enhanced GLP-1 activity it promotes insulin secretion and reduces glucagon release Animal model studies including experiments in obese Zucker rats demonstrated improved glucose tolerance increased beta-cell proliferation decreased apoptosis and neuroprotective effects Vildagliptin is thus relevant for diabetes mellitus type 2 and related biomedical research
Catalog No. 50-000-64048