Learn More
Cayman Chemical VIldaglptIn carylc ACD 5mg
SDP
Supplier:  Cayman Chemical 220885 MG
Encompass_Preferred
A major metabolite of vildagliptin; IC50 = 477 µM for DPP-4 in human Caco-2 cells; formed from hydrolysis of the cyano group on vildagliptin
Catalog No. 50-298-7276