Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villella’s Reagent for Metal Etching 1.9% Picric, 5% HCl in Ethanol, Reagents

Villella’s Reagent for Metal Etching 1.9% Picric, 5% HCl in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$170.00

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Vilellas
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-333-405
Accessories View Documents
Reagents Holdings Llc
CV000200500C
500 mL
Each for $170.00
Only null left
Add to Cart
 
01-333-407
Accessories View Documents
Reagents Holdings Llc
CV000200100C
100 mL N/A N/A N/A
Description

Description

Specifications

Chemical Identifiers

64-17-5 , 7732-18-5 , 67-63-0
46.07
Vilellas
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol

Specifications

Yellow
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
1.9%
Glass Bottle
<1
Liquid
64-17-5 , 7732-18-5 , 67-63-0
Vilellas
CCO
46.07
Reagent
0.82
Non-sterile
Villella’s Reagent for Metal Etching 1.9% Picric, 5% HCl in Ethanol
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.