Learn More
Medchemexpress LLC Viloxazine hydrochloride | 35604-67-2 | MFCD00661100 | 98.9% | 273.75 g/mol | C13H20ClNO3 | 10MM 1ML

Supplier: Medchemexpress LLC HY12578410MM 1ML
Viloxazine hydrochloride is a research-grade small-molecule norepinephrine transporter inhibitor supplied for in vitro pharmacology and biochemical assays. The product is available as solid and as a ready-to-use 10 mM solution in DMSO (1 mL), providing a convenient format for concentration-controlled experiments.
- Ready-to-use 10 mM solution in DMSO, 1 mL
- Also available as solid in multiple mg pack sizes
- High purity (98.9% typical)
- Molecular formula C13H20ClNO3 and molecular weight ~273.75 g/mol
- CAS number 35604-67-2 for unambiguous identification
- SDS and storage instructions provided for safe handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.