Learn More
Medchemexpress LLC Vimirogant hydrochloride | 1802678-42-7 | 98.4% | 589.11 g/mol | C27H36ClF3N4O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY103637A5MG
Encompass_Preferred
Vimirogant hydrochloride (CAS 1802678-42-7) is the hydrochloride salt of vimirogant, a potent and selective RORγt inhibitor used in research to investigate Th17-driven inflammation and autoimmune disease mechanisms. It demonstrates nanomolar activity in biochemical and cellular assays and is supplied at high purity for laboratory use.
  • Potent RORγt inhibitor with reported nanomolar Ki/IC50 values.
  • High purity suitable for research applications.
  • Available in small milligram quantities for preclinical and in vitro studies.
  • Hydrochloride salt form for improved stability and handling.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-46833