Learn More
Medchemexpress LLC Vinburnine | 4880-88-0 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB118010MM/1ML
Encompass_Preferred
Vinburnine, also known as (-)-Eburnamonine or (-)-Vincamone, is a vinca alkaloid and a metabolite of vincamine. It acts as a vasodilator and an allosteric modulator of muscarinic receptors M1-M4. This compound has shown anti-amnesia effects in mouse models and promotes erythrocyte glycolysis, improving tissue oxygen delivery.
  • Acts as a vasodilator
  • Metabolite of vincamine
  • Allosteric modulator of muscarinic receptors M1-M4
  • Decelerates binding of [H]N-methylscopolamine to muscarinic M1-M4 receptors
  • Shows anti-amnesia effect in mouse amnesia models
  • Promotes erythrocyte glycolysis
  • Increases erythrocyte ATP and 2,3-diphosphoglycerol content
  • Improves erythrocyte deformability and tissue oxygen delivery
Catalog No. 50-004-13573