Learn More
Medchemexpress LLC Vincamine | 1617-90-9 | 99.3% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB10211G
Encompass_Preferred
Vincamine is a monoterpenoid indole alkaloid extracted from the Madagascar periwinkle. It acts as a peripheral vasodilator and selectively regulates the brain microcapillary circulation. Vincamine also functions as a GPR40 agonist, protecting β-cells by improving their function and promoting glucose-stimulated insulin secretion (GSIS). It has shown potential in improving glucose homeostasis in vivo for type 2 diabetes mellitus (T2DM) research.
  • Peripheral vasodilator
  • Selective vasoregulator action on brain microcapillary circulation
  • GPR40 agonist
  • Acts as a β-cell protector
  • Ameliorates β-cell dysfunction
  • Promotes glucose-stimulated insulin secretion (GSIS)
  • Improves glucose homeostasis in vivo
  • Potential for type 2 diabetes mellitus (T2DM) research
Catalog No. 50-004-11283