Learn More
Medchemexpress LLC Vincamine | 1617-90-9 | 99.34% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1021100MG
Encompass_Preferred
Vincamine is a monoterpenoid indole alkaloid extracted from the Madagascar periwinkle. It acts as a peripheral vasodilator and selectively regulates brain microcapillary circulation. Vincamine is also a GPR40 agonist, functioning as a β-cell protector by ameliorating β-cell dysfunction and promoting glucose-stimulated insulin secretion (GSIS). It improves glucose homeostasis in vivo and holds potential for research into type 2 diabetes mellitus (T2DM).
  • Peripheral vasodilator
  • Selective vasoregulator action on the brain microcapillary circulation
  • GPR40 agonist
  • β-cell protector by ameliorating β-cell dysfunction
  • Promotes glucose-stimulated insulin secretion (GSIS)
  • Improves glucose homeostasis in vivo
  • Potential for type 2 diabetes mellitus (T2DM) research
Catalog No. 50-004-11281