Learn More
Cayman Chemical VInclozolIn 250mg
SDP
Supplier:  Cayman Chemical 23939250 MG
Encompass_Preferred
A dicarboximide fungicide; active against the plant pathogenic fungi S. sclerotiorum and N. crassa (EC50s = 0.18 and 1.4 UG/ml, respectively); decreases the incidence of leaf drop in lettuce fields experimentally infected with S. sclerotiorum at 0.56 or 1.12 kg/ha
Catalog No. 50-299-5036