Learn More
Cayman Chemical VIncrIstIn-d3 1mg
SDP
Supplier:  Cayman Chemical 345121 MG
Encompass_Preferred
An internal standard for the quantification of vincristine by GC- or LC-MS
Catalog No. 50-294-8040