Learn More
Medchemexpress LLC Vincristine-d3-ester sulfate | 1217854-24-4 | 926.05 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0488S11MG
Encompass_Preferred
Vincristine-d3-ester (sulfate) is a deuterium-labeled Vincristine sulfate, an antitumor vinca alkaloid that inhibits microtubule formation in mitotic spindles. This action causes dividing cells to arrest at the metaphase stage by binding to microtubules with a Ki of 85 nM. Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Inhibits microtubule formation in mitotic spindles.
  • Leads to metaphase arrest in dividing cells.
  • Used in research to study drug pharmacokinetics and metabolism.
Catalog No. 50-003-88589