Learn More
Medchemexpress LLC Vincristine sulfate | 2068-78-2 | 923.04 | 1 ML
Supplier: Medchemexpress LLC HYN048810MM/1ML
Vincristine sulfate is an antitumor vinca alkaloid that inhibits microtubule formation in mitotic spindles, leading to an arrest of dividing cells at the metaphase stage. It binds to microtubules with a Ki of 85 nM. This product is for research use only.
- Inhibits microtubule formation
- Causes metaphase arrest in dividing cells
- Induces apoptosis in tumor cells
- Research use only