Learn More
Enzo Life Sciences Vincristine. sulfate (5 mg). CAS: 2068-78-2
SDP
Supplier:  Enzo Life Sciences BMLT1170005
Encompass_Preferred
Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Depolymerizes microtubules and blocks binding of tubulin to microtubule proteins. Causes aberrant tubulin polymerization. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent. Posttranscriptionally downregulates HIF-1α expression. Alternative name: VCR . sulfate, Leukocristine . sulfate. Purity: ≥98% TLC. Solubility: Soluble in water (25mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-2210