Learn More
Medchemexpress LLC Vindesine sulfate | 59917-39-4 | 99.4% | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY12907110MM/1ML
Encompass_Preferred
Vindesine sulfate is a potent tubulin inhibitor that demonstrates anti-proliferation effects in vitro and antitumor effects in vivo. It effectively inhibits L-cells growth, showing approximately 25% inhibition at 40 nM. This product is suitable for various research applications involving cellular growth and tubulin inhibition.
  • Potent tubulin inhibitor with a Ki of 0.110 μM
  • Exhibits anti-proliferation effects in vitro
  • Demonstrates antitumor effects in vivo
  • Inhibits L-cells growth at 40 nM
Catalog No. 50-003-39331