Learn More
Apexbio Technology LLC Vinorelbine ditartrate, 25mg. Cas: 125317-39-7 MFCD: MFCD03613607. Anti-mitotic chemotherapy drug
SDP
Supplier:  Apexbio Technology LLC A392125
Encompass_Preferred
Description: IC50 Value: 1.25 nM (Hela cell) [1] Vinorelbine is an anti-mitotic chemotherapy drug that is given as a treatment for some types of cancer, including breast cancer and non-small cell lung cancer. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4847