Learn More
Cayman Chemical VInorelbIntartrate 5mg
SDP
Supplier:  Cayman Chemical 212625 MG
Encompass_Preferred
An inhibitor of microtubule polymerization that blocks mitosis at the metaphase/anaphase transition, preventing HeLa cell proliferation (IC50 = 1.25 nM)
Catalog No. 50-298-5406