Learn More
Cayman Chemical VInyl-L-NIOhydrochlorIde 100mg
SDP
Supplier:  Cayman Chemical 80330100 MG
Encompass_Preferred
A potent, selective inhibitor of nNOS with Ki values for inhibition of nNOS, eNOS and iNOS of 100 nM, 12 µM and 60 µM, respectively
Catalog No. 50-289-4229