Learn More
Medchemexpress LLC Violanthin | 40581-17-7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN689510MG
Encompass_Preferred
Violanthin is isolated from the stems of Dendrobium officinale. It has potent antioxidant and antibacterial activities, and inhibits acetylcholinesterase (AChE) with an IC50 value of 79.80 μM.
  • Isolated from the stems of Dendrobium officinale
  • Potent antioxidant activity
  • Potent antibacterial activity
  • Inhibits acetylcholinesterase (AChE)
Catalog No. 50-004-19824