Learn More
Medchemexpress LLC Violanthin | 40581-17-7 | MFCD29037226 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN68951MG
Encompass_Preferred
Violanthin is isolated from the stems of *Dendrobium officinale* and possesses potent antioxidant and antibacterial activities. It inhibits acetylcholinesterase (AChE) with an IC50 value of 79.80 μM.
  • Isolated from stems of *Dendrobium officinale*
  • Possesses potent antioxidant activities
  • Possesses antibacterial activities
  • Inhibits acetylcholinesterase (AChE) with an IC50 value of 79.80 μM
Catalog No. 50-004-19501