Learn More
Medchemexpress LLC Violuric acid 5g
SDP
Supplier:  Medchemexpress LLC HYW0970095G
Encompass_Preferred
Violuric acid is a redox mediator used in the laccase system The violuric acid assay is a method to ascertain that the high-redox potential of laccase is not lost during directed evolution[1][2]
Catalog No. 50-002-05158