Learn More
Thermo Scientific Viper Fingertight Fitting Systems, Stainless Steel, 0.13 mm ID
Supplier:  Thermo Scientific 6040.2325
Encompass
Ensure optimal separations and maximize liquid chromatography (LC) performance with virtually zero-dead-volume connections. Thermo Scientific™ Viper™ Fingertight Fitting Systems with stainless steel capillaries are easy to use and enable a fast, tool-free fluidic setup of HPLC and UHPLC systems. These systems are compatible with virtually any column and valve type and suitable for system pressures up to 1310 bar (19,000 psi).
Catalog No. 50-134-7816